Learn More
Sartorius™ 3.2 Standard Flexboy™ with TPE tubes
Designed for the preparation, storage and transport of biopharmaceutical solutions, intermediates and final bulk products.
19265.50 DKK - 24872.90 DKK
Specifications
Disposable | Single-use |
---|---|
For Use With (Application) | preparation, storage and transport of biopharmaceutical solutions, intermediates and final bulk products |
Film Description | S71 |
Port Detail | 3 Ports |
Sterilization Method | Gamma Irradiated |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
15309162
|
Sartorius™
FFB110912 |
20 L |
19265.50 DKK
Each |
Estimated Shipment: 25-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15359162
|
Sartorius™
FFB110913 |
50 L |
24872.90 DKK
Each |
Estimated Shipment: 25-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Single-use, disposable
- Supplied sterilised and ready to use
Specifications
Single-use | |
S71 | |
Gamma Irradiated |
preparation, storage and transport of biopharmaceutical solutions, intermediates and final bulk products | |
3 Ports |