Få mere at vide
3-(2,4,6-Trimethylphenylsulfonyl)-1,2,4-triazole, TRC
Artikelnummer. 30452143
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30452143 1 g
30455672 500 mg
30451608 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30452143 Leverandør Toronto Research Chemicals Leverandørnr. TRCT4823951G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 149591-20-8
Molekylær formel C11 H13 N3 O2 S
Molekylvægt (g/mol) 251.3
Synonym 1H-1,2,4-Triazole, 3-[(2,4,6-trimethylphenyl)sulfonyl]- (9CI), 5-[(2,4,6-Trimethylphenyl)sulfonyl]-1H-1,2,4-triazole, 3-(2,4,6-Trimethylbenzenesulfonyl)-1,2,4-triazole, 3-(2,4,6-Trimethylphenylsulfonyl)-1,2,4-triazole
IUPAC navn 3-(2,4,6-trimethylphenyl)sulfonyl-4H-1,2,4-triazole
SMIL Cc1cc(C)c(c(C)c1)S(=O)(=O)c2nnc[nH]2
Tekniske data
Kemisk navn eller materiale 3-(2,4,6-Trimethylphenylsulfonyl)-1,2,4-triazole
Mængde 1 g
InChI formel InChI=1S/C11H13N3O2S/c1-7-4-8(2)10(9(3)5-7)17(15,16)11-12-6-13-14-11/h4-6H,1-3H3,(H,12,13,14)
Formel vægt 251.0728