Få mere at vide
(2S)-4'-Depropyl-4'-propylidenelincomycin (Mixture of Diastereomers), TRC
Artikelnummer. 30357976
Skift visning
Mængde:
0.5 mg
5 mg
Pakningsstørrelse:
0.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30357976 5 mg -
30350477 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30357976 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2819155MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 37744-65-3
Molekylær formel C18H32N2O6S
Molekylvægt (g/mol) 404.52
Synonym Lincomycin EP Impurity B, Methyl 6,8-Dideoxy-6-[[[(2S)-1-methyl-4-propylidene-2-pyrrolidinyl]carbonyl]amino]-1-thio-D-erythro-alpha-D-galacto-octopyranoside, Methyl (2S)-6,8-Dideoxy-6-[[(1-methyl-4-propylidene-2-pyrrolidinyl)carbonyl]amino]-1-thio-D-erythro-alpha-D-galacto-octopyranoside, (S)-N-((1R,2R)-2-Hydroxy-1-((2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(methylthio)tetrahydro-2H-pyran-2-yl)propyl)-1-methyl-4-propylidenepyrrolidine-2-carboxamide, 4'-Depropyl-4'-propylidenelincomycin
SMIL CCC=C1C[C@H](N(C)C1)C(=O)N[C@H]([C@@H](C)O)[C@H]2O[C@H](SC)[C@H](O)[C@@H](O)[C@H]2O
Tekniske data
Kemisk navn eller materiale (2S)-4'-Depropyl-4'-propylidenelincomycin (Mixture of Diastereomers)
Mængde 5 mg
InChI formel InChI=1S/C18H32N2O6S/c1-5-6-10-7-11(20(3)8-10)17(25)19-12(9(2)21)16-14(23)13(22)15(24)18(26-16)27-4/h6,9,11-16,18,21-24H,5,7-8H2,1-4H3,(H,19,25)/t9-,11+,12-,13+,14-,15-,16-,18-/m1/s1
Formel vægt 404.1981