Få mere at vide
(2E,4Z)-2,4-Decadienoic Acid Methyl Ester (98%), TRC
Artikelnummer. 30134274
Skift visning
Mængde:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134274 10 g
30007725 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30134274 Leverandør Toronto Research Chemicals Leverandørnr. TRCD21060010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 4493-42-9
Molekylær formel C11 H18 O2
Molekylvægt (g/mol) 182.26
Synonym 2,4-Decadienoic acid, methyl ester, (2E,4Z)-, 2,4-Decadienoic acid, methyl ester, (E,Z)- (8CI), (2E,4Z)-2,4-Decadienoic acid methyl ester, Methyl (2E,4Z)-2,4-decadienoate, Methyl (2E,4Z)-2,4-decadienoate, Methyl (E,Z)-2,4-decadienoate, Methyl 2-trans-4-cis-decadienoate, Methyl 2E,4Z-decadienoate, Methyl trans-2,cis-4-decadienoate
IUPAC navn methyl (2E,4Z)-deca-2,4-dienoate
SMIL CCCCC\C=C/C=C/C(=O)OC
Tekniske data
Kemisk navn eller materiale (2E,4Z)-2,4-Decadienoic Acid Methyl Ester (98%)
Mængde 10 g
InChI formel InChI=1S/C11H18O2/c1-3-4-5-6-7-8-9-10-11(12)13-2/h7-10H,3-6H2,1-2H3/b8-7-,10-9+
Formel vægt 182.1307