Få mere at vide
21-Dehydro Budesonide, TRC
Artikelnummer. 17901485
Skift visning
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17901485 50 mg
17912635 25 mg
17994355 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17901485 Leverandør Toronto Research Chemicals Leverandørnr. TRCD22930550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 85234-63-5
Molekylær formel C25 H32 O6
Molekylvægt (g/mol) 428.52
Synonym (11β,16α)-16,17-[Butylidenebis(oxy)]-11-hydroxy-3,20-dioxo-pregna-1,4-dien-21-al, 16α,17-[(1RS)-Butylidenebis(oxy)]-11β-hydroxy-3,20-dioxopregna-1,4-dien-21-al (21-Dehydrobudesonide), Budesonide Imp. D (EP)
SMIL CCCC1O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)C=O
Tekniske data
Kemisk navn eller materiale 21-Dehydro Budesonide
Renhedsgrad noter HPLC
Mængde 50 mg
InChI formel InChI=1S/C25H32O6/c1-4-5-21-30-20-11-17-16-7-6-14-10-15(27)8-9-23(14,2)22(16)18(28)12-24(17,3)25(20,31-21)19(29)13-26/h8-10,13,16-18,20-22,28H,4-7,11-12H2,1-3H3/t16-,17-,18-,20+,21?,22+,23-,24-,25+/m0/s1
Formel vægt 428.2199
Procent renhed >95%