Få mere at vide
(20R)-17alpha,20-Dihydroxycholesterol, TRC
Artikelnummer. 30010396
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30010396 25 mg
30003440 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30010396 Leverandør Toronto Research Chemicals Leverandørnr. TRCD45018025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 17913-42-7
Molekylær formel C27H46O3
Molekylvægt (g/mol) 418.65
Synonym (3β,20R)-Cholest-5-ene-3,17,20-triol, (20R)-Cholest-5-ene-3β,17,20-triol;
IUPAC navn (3S,8R,9S,10R,13S,14S,17R)-17-[(1R)-1-hydroxy-1,5-dimethyl-hexyl]-10,13-dimethyl-1,2,3,4,7,8,9,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthrene-3,17-diol
SMIL CC(C)CCC[C@@](C)(O)[C@@]1(O)CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Tekniske data
Kemisk navn eller materiale (20R)-17alpha,20-Dihydroxycholesterol
Mængde 25 mg
InChI formel InChI=1S/C27H46O3/c1-18(2)7-6-13-26(5,29)27(30)16-12-23-21-9-8-19-17-20(28)10-14-24(19,3)22(21)11-15-25(23,27)4/h8,18,20-23,28-30H,6-7,9-17H2,1-5H3/t20-,21+,22-,23-,24-,25-,26+,27+/m0/s1
Formel vægt 418.3447