Få mere at vide
20(S)-Hydroxy Prednisolone, TRC
Artikelnummer. 30136430
Skift visning
Mængde:
2.5 mg
25 mg
100 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30136430 100 mg -
30137716 25 mg -
30010840 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136430 Leverandør Toronto Research Chemicals Leverandørnr. TRCH952360100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2299-46-9
Molekylær formel C21 H30 O5
Molekylvægt (g/mol) 362.46
Synonym (20S)-Hydroxyprednisolone
IUPAC navn (8S,9S,10R,11S,13S,14S,17R)-17-[(1S)-1,2-dihydroxyethyl]-11,17-dihydroxy-10,13-dimethyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-one
SMIL C[C@]12C[C@H](O)[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@]2(O)[C@@H](O)CO
Tekniske data
Kemisk navn eller materiale (20S)-Hydroxyprednisolone
Renhedsgrad noter HPLC
Mængde 100 mg
InChI formel InChI=1S/C21H30O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h5,7,9,14-18,22,24-26H,3-4,6,8,10-11H2,1-2H3/t14-,15-,16-,17-,18+,19-,20-,21-/m0/s1
Formel vægt 362.2093
Procent renhed >95%