CAS RN 2299-46-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
(20S)-Hydroxyprednisolone, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
20(S)-Hydroxy Prednisolone, TRC
CAS: 2299-46-9 Molekylær formel: C21 H30 O5 Molekylvægt (g/mol): 362.46 Synonym: (20S)-Hydroxyprednisolone IUPAC navn: (8S,9S,10R,11S,13S,14S,17R)-17-[(1S)-1,2-dihydroxyethyl]-11,17-dihydroxy-10,13-dimethyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-one SMIL: C[C@]12C[C@H](O)[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@]2(O)[C@@H](O)CO