Få mere at vide
[2-(Trimethylammonium)ethyl]methanethiosulfonate Bromide, TRC
Artikelnummer. 30135797
Skift visning
Mængde:
250 mg
2.5 g
Pakningsstørrelse:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30135797 250 mg -
30009282 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30135797 Leverandør Toronto Research Chemicals Leverandørnr. TRCT795900250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 91774-25-3
Molekylær formel C6 H16 N O2 S2 . Br
Molekylvægt (g/mol) 278.23
IUPAC navn trimethyl(2-methylsulfonylsulfanylethyl)azanium;bromide
SMIL [Br-].C[N+](C)(C)CCSS(=O)(=O)C
Tekniske data
Kemisk navn eller materiale (2-(Trimethylammonium)ethyl)methanethiosulfonate Bromide
Mængde 250 mg
InChI formel InChI=1S/C6H16NO2S2.BrH/c1-7(2,3)5-6-10-11(4,8)9;/h5-6H2,1-4H3;1H/q+1;/p-1
Formel vægt 276.9806