Få mere at vide
2-Pyrrolidinyl-3-acetyl Desmorpholinylrocuronium Bromide, TRC
Artikelnummer. 30008406
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30008406 2.5 mg 2.5mg
30006914 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30008406 Leverandør Toronto Research Chemicals Leverandørnr. TRCP9985352.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1190105-66-8
Molekylær formel C34 H55 N2 O4 . Br
Molekylvægt (g/mol) 635.72
Synonym Pyrrolidinium, 1-[(2β,3α,5α,16β,17β)-3,17-bis(acetyloxy)-2-(1-pyrrolidinyl)androstan-16-yl]-1-(2-propen-1-yl)-, bromide (1:1), 1-[3α,17β-Acetoxy-2β-(pyrrolidin-1-yl)-5α-androstan-16β-yl]-1-(prop-2-enyl)pyrrolidinium Bromide
IUPAC navn [(2S,3S,5S,8R,9S,10S,13S,14S,16S,17R)-17-acetyloxy-10,13-dimethyl-16-(1-prop-2-enylpyrrolidin-1-ium-1-yl)-2-pyrrolidin-1-yl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate;bromide
SMIL [Br-].CC(=O)O[C@H]1C[C@@H]2CC[C@@H]3[C@H](CC[C@@]4(C)[C@H]3C[C@@H]([C@@H]4OC(=O)C)[N+]5(CC=C)CCCC5)[C@@]2(C)C[C@@H]1N6CCCC6
Tekniske data
Kemisk navn eller materiale 2-Pyrrolidinyl-3-acetyl Desmorpholinylrocuronium Bromide
Renhedsgrad noter HPLC
Mængde 2.5 mg
InChI formel InChI=1S/C34H55N2O4.BrH/c1-6-17-36(18-9-10-19-36)30-21-28-26-12-11-25-20-31(39-23(2)37)29(35-15-7-8-16-35)22-34(25,5)27(26)13-14-33(28,4)32(30)40-24(3)38;/h6,25-32H,1,7-22H2,2-5H3;1H/q+1;/p-1/t25-,26+,27-,28-,29-,30-,31-,32-,33-,34-;/m0./s1
Formel vægt 634.3345
Procent renhed >95%