Få mere at vide
2-Pyridylacetic Acid-d6 Hydrochloride (d5 Major), TRC
Artikelnummer. 30418659
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30418659 1 mg
30428505 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30418659 Leverandør Toronto Research Chemicals Leverandørnr. TRCP9917521MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1329799-67-8
Molekylær formel C7 2H6 H N O2 . Cl H
Molekylvægt (g/mol) 179.63
Synonym 2,2-Dideuterio-2-(3,4,5,6-tetradeuteriopyridin-2-yl)acetic acid hydrochloride, (2-Pyridinyl)acetic acid D6 hydrochloride, 2-Pyridylacetic acid-d6 hydrochloride, 2-Pyridylacetic-d6 acid hydrochloride
IUPAC navn 2,2-dideuterio-2-(3,4,5,6-tetradeuteriopyridin-2-yl)acetic acid;hydrochloride
SMIL Cl.[2H]c1nc(c([2H])c([2H])c1[2H])C([2H])([2H])C(=O)O
Tekniske data
Kemisk navn eller materiale 2-Pyridylacetic acid-d6 hydrochloride
Renhedsgrad noter HPLC
Mængde 1 mg
InChI formel InChI=1S/C7H7NO2.ClH/c9-7(10)5-6-3-1-2-4-8-6;/h1-4H,5H2,(H,9,10);1H/i1D,2D,3D,4D,5D2;
Formel vægt 179.062
Procent renhed >95%