Få mere at vide
Tocris Bioscience™ 2-Phenylmelatonin
GREENER_CHOICE
Artikelnummer. 18064712
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18064712 10 mg
18035072 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18064712 Leverandør Tocris Bioscience™ Leverandørnr. 0680/10
Potent melatonin agonist
Highly potent melatonin agonist; displays higher affinity and greater potency than melatonin itself. The EC50 values for G protein activation in MT1 and MT2-transfected cells are 65 and 58 pM respectively.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 151889-03-1
Molekylær formel C19H20N2O2
Tekniske data
Kemisk navn eller materiale 2-Phenylmelatonin
Mængde 10 mg
Formel vægt 308.38
Renhed >98%
Molekylvægt (g/mol) 308.381