Få mere at vide
2-Oxo Clopidogrel Carboxylic Acid(Mixture of Diastereomers), TRC
Artikelnummer. 30134291
Skift visning
Mængde:
1 mg
5 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134291 50 mg
30011639 5 mg
30010826 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30134291 Leverandør Toronto Research Chemicals Leverandørnr. TRCO86996050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 109904-36-1
Molekylær formel C15 H14 Cl N O3 S
Molekylvægt (g/mol) 323.79
Synonym α-(2-Chlorophenyl)-2,6,7,7a-tetrahydro-2-oxothieno[3,2-c]pyridine-5(4H)-acetic Acid
IUPAC navn 2-(2-chlorophenyl)-2-(2-oxo-4,6,7,7a-tetrahydrothieno[3,2-c]pyridin-5-yl)acetic acid
SMIL OC(=O)C(N1CCC2SC(=O)C=C2C1)c3ccccc3Cl
Tekniske data
Kemisk navn eller materiale 2-Oxo rac-Clopidogrel carboxylic acid
Mængde 50 mg
InChI formel InChI=1S/C15H14ClNO3S/c16-11-4-2-1-3-10(11)14(15(19)20)17-6-5-12-9(8-17)7-13(18)21-12/h1-4,7,12,14H,5-6,8H2,(H,19,20)
Formel vægt 323.0383