Få mere at vide
2-n-Butylbenzo[b]furan, 98%, Thermo Scientific™
Artikelnummer. 11362057
Skift visning
Mængde:
5 g
25 g
Pakningsstørrelse:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11362057 25 g 25g
11352057 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 11362057 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. L07805.14
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 4265-27-4
Molekylær formel C12H14O
Molekylvægt (g/mol) 174.24
MDL nummer MFCD00005851
InChI nøgle OVJKFJDEVKABNF-UHFFFAOYSA-N
Synonym 2-butylbenzofuran, benzofuran, 2-butyl, 2-n-butylbenzo b furan, 2-butyl-benzofuran, 2-n-butylbenzofuran, 2-butylbenzo b furan, butylbenzofuran, 2-n-butylbezofuran, butyl-2-benzofuran, 2-n-butylbenzofurane
PubChem CID 77951
IUPAC navn 2-butyl-1-benzofuran
SMIL CCCCC1=CC2=CC=CC=C2O1
Tekniske data
Kemisk navn eller materiale 2-n-Butylbenzo[b]furan
Tæthed 0.987
Kogepunkt 114°C to 116°C (8 mmHg)
Flammepunkt 101°C (213°F)
Brydningsindeks 1.535
Mængde 25 g
Beilstein 1365111
Formel vægt 174.24
Procent renhed 98%