Få mere at vide
2-Methylfluoranthene, TRC
Artikelnummer. 30012844
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30012844 25 mg -
30009473 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30012844 Leverandør Toronto Research Chemicals Leverandørnr. TRCM30530525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 33543-31-6
Molekylær formel C17 H12
Molekylvægt (g/mol) 216.28
Synonym Fluoranthene, 2-methyl- (6CI, 7CI, 8CI, 9CI, ACI), 2-Methylfluoranthene (ACI)
IUPAC navn 2-methylfluoranthene
SMIL Cc1cc2c3ccccc3c4cccc(c1)c24
Tekniske data
Kemisk navn eller materiale 2-Methylfluoranthene
Mængde 25 mg
InChI formel InChI=1S/C17H12/c1-11-9-12-5-4-8-15-13-6-2-3-7-14(13)16(10-11)17(12)15/h2-10H,1H3
Formel vægt 216.0939