Få mere at vide
2-(methylamino)benzene-1-sulfonamide, TRC
Artikelnummer. 30481920
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30481920 10 mg
30473463 50 mg
30478542 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30481920 Leverandør Toronto Research Chemicals Leverandørnr. TRCM28890810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 21639-28-1
Molekylær formel C7H10N2O2S
Molekylvægt (g/mol) 186.23
IUPAC navn 2-(methylamino)benzenesulfonamide
SMIL CNc1ccccc1S(=O)(=O)N
Tekniske data
Kemisk navn eller materiale 2-(methylamino)benzene-1-sulfonamide
Mængde 10 mg
InChI formel InChI=1S/C7H10N2O2S/c1-9-6-4-2-3-5-7(6)12(8,10)11/h2-5,9H,1H3,(H2,8,10,11)
Formel vægt 186.0463