missing translation for 'onlineSavingsMsg'
Få mere at vide

2-Methyl-5-nitrobenzenesulfonamide, TRC

Product Code. 30482286
Change view
Click to view available options
Mængde:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30482286 5 g
30471518 25 g
30471519 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30482286 Supplier Toronto Research Chemicals Supplier No. TRCM3211905G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6269-91-6
Molekylær formel C7H8N2O4S
Molekylvægt (g/mol) 216.21
Synonym 5-Nitro-o-toluenesulfonamide, 2-Methyl-5-nitrobenzenesulfonamide, 3-Aminosulfonyl-4-methylnitrobenzene, NSC 33880
IUPAC navn 2-methyl-5-nitrobenzenesulfonamide
SMIL Cc1ccc(cc1S(=O)(=O)N)[N+](=O)[O-]

Specifications

Kemisk navn eller materiale 2-Methyl-5-nitrobenzenesulfonamide
Mængde 5 g
InChI formel InChI=1S/C7H8N2O4S/c1-5-2-3-6(9(10)11)4-7(5)14(8,12)13/h2-4H,1H3,(H2,8,12,13)
Formel vægt 216.0205
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.