Få mere at vide
2-Methyl-5-nitrobenzenesulfonamide, TRC
Artikelnummer. 30471518
Skift visning
Mængde:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30471518 25 g
30471519 1 g
30482286 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30471518 Leverandør Toronto Research Chemicals Leverandørnr. TRCM32119025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 6269-91-6
Molekylær formel C7H8N2O4S
Molekylvægt (g/mol) 216.21
Synonym 5-Nitro-o-toluenesulfonamide, 2-Methyl-5-nitrobenzenesulfonamide, 3-Aminosulfonyl-4-methylnitrobenzene, NSC 33880
IUPAC navn 2-methyl-5-nitrobenzenesulfonamide
SMIL Cc1ccc(cc1S(=O)(=O)N)[N+](=O)[O-]
Tekniske data
Kemisk navn eller materiale 2-Methyl-5-nitrobenzenesulfonamide
Mængde 25 g
InChI formel InChI=1S/C7H8N2O4S/c1-5-2-3-6(9(10)11)4-7(5)14(8,12)13/h2-4H,1H3,(H2,8,12,13)
Formel vægt 216.0205