Få mere at vide
2-Methyl-1,3-propandiol (beta-Hydroxyisobutanol), TRC
Artikelnummer. 30135952
Skift visning
Mængde:
1 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135952 10 g
30135951 50 g
17988014 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30135952 Leverandør Toronto Research Chemicals Leverandørnr. TRCH94363510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2163-42-0
Molekylær formel C4 H10 O2
Molekylvægt (g/mol) 90.121
Synonym β-Hydroxyisobutanol, 1,3-Dihydroxy-2-methylpropane, 1,3-Dihydroxyisobutane, 2-Methyl-1,3-propanediol, 2-Methyl-1,3-propylene Glycol, 2-Methylpropan-1,3-diol, MPDiol, Methylpropanediol
IUPAC navn 2-methylpropane-1,3-diol
SMIL CC(CO)CO
Tekniske data
Kemisk navn eller materiale 2-Methyl-1,3-propandiol
Mængde 10 g
InChI formel InChI=1S/C4H10O2/c1-4(2-5)3-6/h4-6H,2-3H2,1H3
Formel vægt 90.0681