Få mere at vide
2-Methoxyphenol-d3, TRC
Artikelnummer. 30483801
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30483801 250 mg
30477486 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30483801 Leverandør Toronto Research Chemicals Leverandørnr. TRCM265302250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 74495-69-5
Molekylær formel C7 2H3 H5 O2
Molekylvægt (g/mol) 127.16
Synonym Phenol, 2-(methoxy-d3)- (9CI, ACI), 2-(Methoxy-d3)phenol (ACI), 2-(Trideuteriomethoxy)phenol, Guaiacol-d3, 2-Methoxyphenol-D3 (methoxy-D3), 2-Methoxy-d3-phenol, 2-Methoxyphenol-d3
IUPAC navn 2-(trideuteriomethoxy)phenol
SMIL [2H]C([2H])([2H])Oc1ccccc1O
Tekniske data
Kemisk navn eller materiale 2-Methoxyphenol-d3
Mængde 250 mg
InChI formel InChI=1S/C7H8O2/c1-9-7-5-3-2-4-6(7)8/h2-5,8H,1H3/i1D3
Formel vægt 127.0713