CAS RN 74495-69-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
2-Methoxyphenol-d3, TRC
CAS: 74495-69-5 Molekylær formel: C7 2H3 H5 O2 Molekylvægt (g/mol): 127.16 Synonym: Phenol, 2-(methoxy-d3)- (9CI, ACI),2-(Methoxy-d3)phenol (ACI),2-(Trideuteriomethoxy)phenol,Guaiacol-d3,2-Methoxyphenol-D3 (methoxy-D3),2-Methoxy-d3-phenol,2-Methoxyphenol-d3 IUPAC navn: 2-(trideuteriomethoxy)phenol SMIL: [2H]C([2H])([2H])Oc1ccccc1O
2
2-Methoxy-d3-phenol, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.