Få mere at vide
2-Hydroxyethyl Nortadalafil, TRC
Artikelnummer. 30135740
Skift visning
Mængde:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135740 10 mg
30138014 100 mg
30007366 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30135740 Leverandør Toronto Research Chemicals Leverandørnr. TRCH94224510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 385769-94-8
Molekylær formel C23H21N3O5
Molekylvægt (g/mol) 419.43
Synonym 2-Hydroxyethyl Nortadalafil, rel-(6R,12aR)-6-(1,3-Benzodioxol-5-yl)-2,3,6,7,12,12a-hexahydro-2-(2-hydroxyethyl)pyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione
IUPAC navn (2R,8R)-2-(1,3-benzodioxol-5-yl)-6-(2-hydroxyethyl)-3,6,17-triazatetracyclo[8.7.0.0^{3,8}.0^{11,16}]heptadeca-1(10),11,13,15-tetraene-4,7-dione
SMIL OCCN1CC(=O)N2[C@H](Cc3c([nH]c4ccccc34)[C@H]2c5ccc6OCOc6c5)C1=O
Tekniske data
Kemisk navn eller materiale 2-Hydroxyethyl Nortadalafil
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/C23H21N3O5/c27-8-7-25-11-20(28)26-17(23(25)29)10-15-14-3-1-2-4-16(14)24-21(15)22(26)13-5-6-18-19(9-13)31-12-30-18/h1-6,9,17,22,24,27H,7-8,10-12H2/t17-,22-/m1/s1
Formel vægt 419.1481
Procent renhed >95%