Få mere at vide
2-Fluoro-N-phenylbenzamide, 97%, Thermo Scientific™
Artikelnummer. 15465248
Skift visning
Mængde:
250 mg
1 g
Pakningsstørrelse:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
15465248 250 mg 250mg
15455248 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 15465248 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. H59063.MD
Request Bulk or Custom Format
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1747-80-4
Molekylær formel C13H10FNO
Molekylvægt (g/mol) 215.227
MDL nummer MFCD00454563
InChI nøgle LFYRRNCQLJGEGP-UHFFFAOYSA-N
Synonym 2-fluorobenzanilide, 2-fluoro-n-phenyl-benzamide, 2-fluorophenyl-n-benzamide, benzamide,2-fluoro-n-phenyl
PubChem CID 242890
IUPAC navn 2-fluoro-N-phenylbenzamide
SMIL C1=CC=C(C=C1)NC(=O)C2=CC=CC=C2F
Tekniske data
Kemisk navn eller materiale 2-Fluoro-N-phenylbenzamide
Mængde 250 mg
Formel vægt 215.23
Procent renhed 97%
Fysisk form Solid