Få mere at vide
2-(Diphenylmethoxy)-N-ethylamine, TRC
Artikelnummer. 30376579
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30376579 100 mg
30370042 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30376579 Leverandør Toronto Research Chemicals Leverandørnr. TRCD491755100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 17349-94-9
Molekylær formel C15H17NO
Molekylvægt (g/mol) 227.3
Synonym 2-(Diphenylmethoxy)ethylamine, Dinordiphenhydramine, FAR 9058D, N,N-Didemethyldiphenhydramine, NSC 55993
SMIL C1=CC=C(C=C1)C(C2=CC=CC=C2)OCCN
Tekniske data
Kemisk navn eller materiale 2-(Diphenylmethoxy)-N-ethylamine
Mængde 100 mg
InChI formel InChI=1S/C15H17NO/c16-11-12-17-15(13-7-3-1-4-8-13)14-9-5-2-6-10-14/h1-10,15H,11-12,16H2