Få mere at vide
2-(Dibenzylamino)-3',4'-dihydroxy-acetophenone, TRC
Artikelnummer. 30134194
Skift visning
Mængde:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134194 25 mg
30133283 100 mg
17971185 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30134194 Leverandør Toronto Research Chemicals Leverandørnr. TRCD41752025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 13062-58-3
Molekylær formel C22H21NO3
Molekylvægt (g/mol) 347.41
Synonym N,N-Dibenzylarterenone, 2-[Bis(phenylmethyl)amino]-1-(3,4-dihydroxyphenyl)-ethanone;
IUPAC navn 2-(dibenzylamino)-1-(3,4-dihydroxyphenyl)ethanone
SMIL OC1=C(O)C=CC(C(CN(CC2=CC=CC=C2)CC3=CC=CC=C3)=O)=C1
Tekniske data
Kemisk navn eller materiale 2-(Dibenzylamino)-3',4'-dihydroxy-acetophenone
Mængde 25 mg
InChI formel InChI=1S/C22H21NO3/c24-20-12-11-19(13-21(20)25)22(26)16-23(14-17-7-3-1-4-8-17)15-18-9-5-2-6-10-18/h1-13,24-25H,14-16H2
Formel vægt 347.1521