Få mere at vide
2-Deoxy-2,2-difluoro-D-erythro-pentofuranos-1-ulose-3,5-dibenzoate, TRC
Artikelnummer. 30363141
Skift visning
Mængde:
1 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30363141 100 g
30367464 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30363141 Leverandør Toronto Research Chemicals Leverandørnr. TRCD232775100G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 122111-01-7
Molekylær formel C19 H14 F2 O6
Molekylvægt (g/mol) 376.31
Synonym D-erythro-Pentonic acid, 2-deoxy-2,2-difluoro-, γ-lactone, 3,5-dibenzoate
IUPAC navn [(2R,3R)-3-benzoyloxy-4,4-difluoro-5-oxooxolan-2-yl]methyl benzoate
SMIL FC1(F)[C@H](OC(=O)c2ccccc2)[C@@H](COC(=O)c3ccccc3)OC1=O
Tekniske data
Kemisk navn eller materiale 2-Deoxy-2,2-difluoro-D-erythro-pentofuranos-1-ulose-3,5-dibe
Mængde 100 g
InChI formel InChI=1S/C19H14F2O6/c20-19(21)15(27-17(23)13-9-5-2-6-10-13)14(26-18(19)24)11-25-16(22)12-7-3-1-4-8-12/h1-10,14-15H,11H2/t14-,15-/m1/s1
Formel vægt 376.0758