Få mere at vide
2-Cyclohexyl-2-hydroxy-phenylacetic Acid, TRC
Artikelnummer. 30133808
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133808 5 g
17933835 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30133808 Leverandør Toronto Research Chemicals Leverandørnr. TRCC9879255G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 4335-77-7
Molekylær formel C14 H18 O3
Molekylvægt (g/mol) 234.29
Synonym (RS)-2-Cyclohexyl-2-hydroxy-2-phenylacetic Acid, Oxybutynin Hydrochloride Imp. D (EP), Phenylcyclohexylglycolic Acid
IUPAC navn 2-cyclohexyl-2-hydroxy-2-phenylacetic acid
SMIL OC(=O)C(O)(C1CCCCC1)c2ccccc2
Tekniske data
Kemisk navn eller materiale Phenylcyclohexylglycolic Acid
Mængde 5 g
InChI formel InChI=1S/C14H18O3/c15-13(16)14(17,11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1,3-4,7-8,12,17H,2,5-6,9-10H2,(H,15,16)
Formel vægt 234.1256