Få mere at vide
2-Chloro-N-5-quinolinylacetamide, TRC
Artikelnummer. 30350520
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30350520 10 mg
30365330 100 mg
30361637 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30350520 Leverandør Toronto Research Chemicals Leverandørnr. TRCC36943810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 121221-08-7
Molekylær formel C11H9N2OCl
Molekylvægt (g/mol) 220.65
IUPAC navn 2-chloro-N-quinolin-5-ylacetamide
SMIL ClCC(=O)Nc1cccc2ncccc12
Tekniske data
Kemisk navn eller materiale 2-Chloro-N-5-quinolinylacetamide
Mængde 10 mg
InChI formel InChI=1S/C11H9ClN2O/c12-7-11(15)14-10-5-1-4-9-8(10)3-2-6-13-9/h1-6H,7H2,(H,14,15)
Formel vægt 220.0403