Få mere at vide
2-Chloro Hippuric Acid, TRC
Artikelnummer. 30362663
Skift visning
Mængde:
250 mg
1 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30362663 1 g
30349696 10 g
30350269 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30362663 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3665451G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 16555-60-5
Molekylær formel C9H8ClNO3
Molekylvægt (g/mol) 213.62
Synonym (2-Chlorobenzoylamino)acetic Acid, N-(2-Chlorobenzoyl)glycine, N-(o-Chlorobenzoyl)glycine, o-Chlorobenzoylglycine, o-Chlorohippuric Acid
IUPAC navn 2-[(2-chlorobenzoyl)amino]acetic acid
SMIL OC(=O)CNC(=O)c1ccccc1Cl
Tekniske data
Kemisk navn eller materiale 2-Chloro Hippuric Acid
Mængde 1 g
InChI formel InChI=1S/C9H8ClNO3/c10-7-4-2-1-3-6(7)9(14)11-5-8(12)13/h1-4H,5H2,(H,11,14)(H,12,13)
Formel vægt 213.0193