Få mere at vide
2-Chloro-4-nitrophenyl Alpha-D-Maltotrioside, TRC
Artikelnummer. 30137492
Skift visning
Mængde:
25 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137492 500 mg
30136017 25 mg
17928304 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137492 Leverandør Toronto Research Chemicals Leverandørnr. TRCC596473500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 118291-90-0
Molekylær formel C24H34ClNO18
Molekylvægt (g/mol) 659.98
Synonym 2-Chloro-4-nitrophenyl alpha-Maltotrioside, 2-Chloro-4-nitrophenyl O-alpha-D-glucopyranosyl-(1-4)-O-alpha-D-glucopyranosyl-(1-4)-alpha-D-glucopyranoside
IUPAC navn (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-[(2R,3S,4R,5R,6R)-6-(2-chloro-4-nitrophenoxy)-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMIL OC[C@H]1OC(O[C@H]2C(O)[C@@H](O)C(O[C@H]3C(O)[C@@H](O)C(Oc4ccc(cc4Cl)[N+](=O)[O-])O[C@@H]3CO)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O
Tekniske data
Kemisk navn eller materiale 2-Chloro-4-nitrophenyl alpha-D-Maltotrioside
Mængde 500 mg
InChI formel InChI=1S/C24H34ClNO18/c25-8-3-7(26(37)38)1-2-9(8)39-22-18(35)15(32)20(11(5-28)41-22)44-24-19(36)16(33)21(12(6-29)42-24)43-23-17(34)14(31)13(30)10(4-27)40-23/h1-3,10-24,27-36H,4-6H2/t10-,11-,12-,13-,14+,15-,16-,17-,18-,19-,20-,21-,22+,23-,24-/m1/s1
Formel vægt 659.1464