missing translation for 'onlineSavingsMsg'
Få mere at vide

2-Chloro-3,5-dinitrobenzotrifluoride, 98+%

Artikelnummer. 11448247
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 g
25 g
Pakningsstørrelse:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde Pakningsstørrelse
11448247 25 g 25g
11438247 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 11448247 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. A17617.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

CAS: 392-95-0 | C7H2ClF3N2O4 | 270.55 g/mol

It is used to increase mutagenic activity in the glutathione transferase mutant (C36).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to increase mutagenic activity in the glutathione transferase mutant (C36).

Solubility
Insoluble in water. Soluble in Chloroform, Methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed in a cool, dry place. Store under 20˚C Freezer, Under Inert Atmosphere.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 392-95-0
Molekylær formel C7H2ClF3N2O4
Molekylvægt (g/mol) 270.55
MDL nummer MFCD00007076
InChI nøgle RLXKADBMLQPLDV-UHFFFAOYSA-N
Synonym 2-chloro-3,5-dinitrobenzotrifluoride, 2-chloro-1,5-dinitro-3-trifluoromethyl benzene, benzene, 2-chloro-1,5-dinitro-3-trifluoromethyl, 2-chloro-3,5-binitrotrifluorotoluene, pubchem4846, 2-chlor-1,5-dinitro-3-trifluormethyl benzol, ksc914e2d, labotest-bb lt00159813, labotest-bb lt03380099, 2-chloro-3,5-dinitro-benzotrifluoride
PubChem CID 67851
IUPAC navn 2-chloro-1,5-dinitro-3-(trifluoromethyl)benzene
SMIL [O-][N+](=O)C1=CC(=C(Cl)C(=C1)C(F)(F)F)[N+]([O-])=O

Tekniske data

Kemisk navn eller materiale 2-Chloro-3,5-dinitrobenzotrifluoride
Smeltepunkt 61°C to 64°C
Mængde 25 g
FN nummer UN2923
Beilstein 2061156
Opløselighedsinformation Insoluble in water. Soluble in Chloroform,Methanol.
Formel vægt 270.55
Procent renhed ≥98%

RUO– Kun brug til forskning

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.