missing translation for 'onlineSavingsMsg'
Få mere at vide

2-chloro-1H-indole-3-carbaldehyde, Maybridge

Artikelnummer. 10097929
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
Emballage:
Amber Glass Bottle
Pakningsstørrelse:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde Emballage unitSize
10097929 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 10097929 Leverandør Thermo Scientific Maybridge Leverandørnr. MAY00304DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy

Tekniske data

Kemisk navn eller materiale 2-chloro-1H-indole-3-carbaldehyde
CAS 5059-30-3
Emballage Amber Glass Bottle
Molekylær formel C9H6ClNO
Mængde 1 g
Synonym 2-chloro-indole-3-carbaldehyde, 2-chloroindole-3-carbaldehyde, 2-chloro-1h-indole-3-carboxaldehyde, 2-chloroindole-3-carboxaldehyde, 2-chloro-3-formyl-1h-indole, 1h-indole-3-carboxaldehyde, 2-chloro, chloroindolecarbaldehyde, 2-chloro-3-formylindol, 1h-indole-3-carboxaldehyde,2-chloro
InChI nøgle XYSSNBNFOBVMAU-UHFFFAOYSA-N
SMIL C1=CC=C2C(=C1)C(=C(N2)Cl)C=O
IUPAC navn 2-chloro-1H-indole-3-carbaldehyde
Molekylvægt (g/mol) 179.603
PubChem CID 314791
Formel vægt 179.61
Procent renhed 95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.