Få mere at vide
2-Bromo-5-chlorotoluene, 98%, Thermo Scientific™
Artikelnummer. 11359669
Skift visning
Mængde:
25 g
50 g
250 g
Pakningsstørrelse:
250g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde Pakningsstørrelse
11359669 25 g 25g
11478096 50 g 50g
11488096 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 11359669 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. A14132.14
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 14495-51-3
Molekylær formel C7H6BrCl
Molekylvægt (g/mol) 205.479
MDL nummer MFCD00000602
InChI nøgle RTIPTGMVQIIMKL-UHFFFAOYSA-N
Synonym 2-bromo-5-chlorotoluene, benzene, 1-bromo-4-chloro-2-methyl, toluene, 2-bromo-5-chloro, 6-bromo-3-chlorotoluene, 1-bromo-4-chloro-2-methyl-benzene, 5-chloro-2-bromotoluene, pubchem3600, acmc-209ctq, 2-bromo-5-chloro-toluene, ksc493i7b
PubChem CID 84476
IUPAC navn 1-bromo-4-chloro-2-methylbenzene
SMIL CC1=C(C=CC(=C1)Cl)Br
Tekniske data
Kemisk navn eller materiale 2-Bromo-5-chlorotoluene
Tæthed 1.545
Mængde 25 g
Lugt Aromatic
Brydningsindeks 1.575
Beilstein 2500157
Kogepunkt 98°C to 99°C (25 mmHg)
Formel vægt 205.48
Procent renhed 98%