Få mere at vide
2-Aminodipyrido[1,2-a:3',2’-d]imidazole Dihydrochloride, TRC
Artikelnummer. 30132653
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132653 50 mg
30004333 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30132653 Leverandør Toronto Research Chemicals Leverandørnr. TRCA60880150MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1980075-70-4
Molekylær formel C10H10Cl2N4
Molekylvægt (g/mol) 257.12
Synonym Pyrido[3',2':4,5]imidazo[1,2-a]pyridin-2-amine Dihydrochloride, Dipyrido[1,2-a:3',2'-d]imidazol-2-amine Dihydrochloride, Glu-P-2;
IUPAC navn 1,3,8-triazatricyclo[7.4.0.02,7]trideca-2(7),3,5,8,10,12-hexaen-4-amine;dihydrochloride
SMIL NC1=NC(N2C(C=CC=C2)=N3)=C3C=C1.[2HCl]
Tekniske data
Kemisk navn eller materiale 2-Aminodipyrido[1,2-a:3',2'-d]imidazole Dihydrochloride
Analyt- eller komponentnavne 2-Aminodipyrido[1,2-a:3',2'-d]imidazole Dihydrochloride
Renhedsgrad noter HPLC
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C10H8N4.2ClH/c11-8-5-4-7-10(13-8)14-6-2-1-3-9(14)12-7;;/h1-6H,(H2,11,13);2*1H
Løsningstype Neat
Industry Type Forensics
Mængde 50 mg
Procent renhed >95%