Få mere at vide
2-Amino-N-cyclohexyl-N-methylbenzylamine, TRC
Artikelnummer. 17942235
Skift visning
Mængde:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17942235 250 mg
17933935 1 g
17941265 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17942235 Leverandør Toronto Research Chemicals Leverandørnr. TRCA603680250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 57365-08-9
Molekylvægt (g/mol) 218.34
SMIL CN(Cc1ccccc1N)C2CCCCC2
Tekniske data
Kemisk navn eller materiale 2-Amino-N-cyclohexyl-N-methylbenzylamine
Renhedsgrad noter HPLC
Molekylær formel C14H22N2
Mængde 250 mg
Synonym Bromhexine Hydrochloride Imp. C (EP), Bromhexine Imp. C (EP), N-(2-Aminobenzyl)-N-methylcyclohexanamine, Bromhexine Hydrochloride Impurity C, Bromhexine Impurity C
Formel vægt 218.1783
Procent renhed >95%