Få mere at vide
(2-Amino-1H-benzimidazol-6-yl)(4-fluorophenyl)-methanone, TRC
Artikelnummer. 30133972
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133972 50 mg
30004991 5 mg
30011737 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133972 Leverandør Toronto Research Chemicals Leverandørnr. TRCA60153550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 82050-13-3
Molekylær formel C14 H10 F N3 O
Molekylvægt (g/mol) 255.25
Synonym Methanone, (2-amino-1H-benzimidazol-5-yl)(4-fluorophenyl)- (9CI), (2-Amino-1H-benzimidazol-6-yl)(4-fluorophenyl)methanone, 2-Aminoflubendazole, Hydrolyzed-flubendazole
IUPAC navn (2-amino-1H-benzimidazol-5-yl)-(4-fluorophenyl)methanone
SMIL Nc1nc2cc(ccc2[nH]1)C(=O)c3ccc(F)cc3
Tekniske data
Kemisk navn eller materiale 2-Aminoflubendazole
Mængde 50 mg
InChI formel InChI=1S/C14H10FN3O/c15-10-4-1-8(2-5-10)13(19)9-3-6-11-12(7-9)18-14(16)17-11/h1-7H,(H3,16,17,18)
Formel vægt 255.0808