missing translation for 'onlineSavingsMsg'
Få mere at vide

2,6-Difluorobenzyl alcohol, 97%, Thermo Scientific™

Artikelnummer. 11413907
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 g
25 g
Pakningsstørrelse:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11413907 25 g 25g
11403907 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 11413907 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. A18336.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2,6-Difluorobenzyl alcohol is used to get 2-(bromomethyl)-1,3-difluorobenzene.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 19064-18-7
Molekylær formel C7H6F2O
Molekylvægt (g/mol) 144.121
MDL nummer MFCD00004603
InChI nøgle LVICICZQETYOGS-UHFFFAOYSA-N
Synonym 2,6-difluorobenzyl alcohol, 2,6-difluorophenyl methanol, benzenemethanol, 2,6-difluoro, 2,6-difluorobenzylalcohol, 2,6-difluorobenzenemethanol, 2,6-difluoro-phenyl-methanol, 2,6-difluoro benzyl alcohol, 2,6-difluorophenyl methan-1-ol, pubchem4914, acmc-209etz
PubChem CID 87921
IUPAC navn (2,6-difluorophenyl)methanol
SMIL C1=CC(=C(C(=C1)F)CO)F

Tekniske data

Kemisk navn eller materiale 2,6-Difluorobenzyl alcohol
Tæthed 1.3
Kogepunkt 87°C to 88°C (14 mmHg)
Flammepunkt 88°C (190°F)
Brydningsindeks 1.495
Mængde 25 g
Beilstein 2501536
Opløselighedsinformation Not miscible or difficult to mix in water.
Formel vægt 144.12
Procent renhed 97%
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.