missing translation for 'onlineSavingsMsg'
Få mere at vide

2,6-Dibromo-4-(trifluoromethoxy)aniline, 97%, Thermo Scientific™

Artikelnummer. 15422058
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
5 g
Pakningsstørrelse:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde Pakningsstørrelse
15422058 1 g 1g
15432058 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15422058 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. H55725.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88149-49-9
Molekylær formel C7H4Br2F3NO
Molekylvægt (g/mol) 334.92
MDL nummer MFCD00153113
InChI nøgle JBSWOEGXMADXOU-UHFFFAOYSA-N
Synonym 2,6-dibromo-4-trifluoromethoxy aniline, bromo riluzole impurity, 3,5-dibromo-4-aminotrifluoromethoxybenzene, 2,6-dibromo-4-trifluoromethoxy benzenamine, 2-amino-1,3-dibromo-5-trifluoromethoxy benzene, 2,6dibromo-4-trifluoromethoxy aniline, 2,6-dibromo-4-trifluoro-methoxy aniline, benzenamine, 2,6-dibromo-4-trifluoromethoxy, pubchem10031
PubChem CID 2736797
IUPAC navn 2,6-dibromo-4-(trifluoromethoxy)aniline
SMIL NC1=C(Br)C=C(OC(F)(F)F)C=C1Br

Specifications

Kemisk navn eller materiale 2,6-Dibromo-4-(trifluoromethoxy)aniline
Smeltepunkt 70°C to 74°C
Mængde 1 g
Formel vægt 334.92
Procent renhed 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.