Learn More
JT Baker 2,5LT Formic Acid, 98% BAKER ANALYZED Reagent
Brand: JT Baker 6037.2500
2,5LT Formic Acid, 98% BAKER ANALYZED Reagent
Safety and Handling
- Danger
Hazard Category
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H226-Flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.