Få mere at vide
2-(4-Methoxybenzyl)isothiazolidine 1,1-Dioxide, TRC
Artikelnummer. 30471869
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30471869 50 mg
30469418 10 mg
30464718 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30471869 Leverandør Toronto Research Chemicals Leverandørnr. TRCM26590350MG
Denne vare kan ikke returneres. Se returpolitik
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 158089-76-0
Molekylær formel C11 H15 N O3 S
Molekylvægt (g/mol) 241.3067
Synonym Isothiazolidine, 2-[(4-methoxyphenyl)methyl]-, 1,1-dioxide
IUPAC navn 2-[(4-methoxyphenyl)methyl]-1,2-thiazolidine 1,1-dioxide
SMIL COc1ccc(CN2CCCS2(=O)=O)cc1
Tekniske data
Kemisk navn eller materiale N-(4-Methoxybenzyl)-1,3-propanesultam
Mængde 50 mg
InChI formel InChI=1S/C11H15NO3S/c1-15-11-5-3-10(4-6-11)9-12-7-2-8-16(12,13)14/h3-6H,2,7-9H2,1H3
Formel vægt 241.0773