Få mere at vide
2,4-Disulfamyl-5-trifluoromethylaniline, TRC
Artikelnummer. 30134616
Skift visning
Mængde:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134616 5 g
30135441 10 g
30008334 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30134616 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4935805G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 654-62-6
Molekylær formel C7 H8 F3 N3 O4 S2
Molekylvægt (g/mol) 319.28
Synonym 4-Amino-6-(trifluoromethyl)benzene-1,3-disulphonamide, 1,3-Benzenedisulfonamide, 4-amino-6-(trifluoromethyl)-, Toluene-2,4-disulfonamide, 5-amino-α,α,α-trifluoro- (6CI,7CI,8CI), 2,4-Disulfamoyl-5-trifluoromethylaniline, 4-Amino-6-(trifluoromethyl)-1,3-benzenedisulfonamide, 5-Trifluoromethyl-2,4-disulfamoylaniline, NSC 44625
IUPAC navn 4-amino-6-(trifluoromethyl)benzene-1,3-disulfonamide
SMIL Nc1cc(c(cc1S(=O)(=O)N)S(=O)(=O)N)C(F)(F)F
Tekniske data
Kemisk navn eller materiale 4-amino-6-(trifluoromethyl)benzene-1,3-disulfonamide
Mængde 5 g
InChI formel InChI=1S/C7H8F3N3O4S2/c8-7(9,10)3-1-4(11)6(19(13,16)17)2-5(3)18(12,14)15/h1-2H,11H2,(H2,12,14,15)(H2,13,16,17)
Formel vægt 318.9908