missing translation for 'onlineSavingsMsg'
Få mere at vide

2,4-Dimethyl-2-pentanol, 96%, Thermo Scientific™

Artikelnummer. 11312716
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
5 g
Pakningsstørrelse:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde Pakningsstørrelse
11312716 5 g 5g
11302716 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 11312716 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. L08795.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-06-9
Molekylær formel C7H16O
Molekylvægt (g/mol) 116.20
MDL nummer MFCD00021811
InChI nøgle FMLSQAUAAGVTJO-UHFFFAOYSA-N
Synonym 2,4-dimethyl-2-pentanol, isobutyldimethylcarbinol, 2-pentanol, 2,4-dimethyl, acmc-20aoch, 2-pentanol,4-dimethyl, 2-pentanol,2,4-dimethyl, 2-pentanol, 2,4-dimethyl-8ci 9ci
PubChem CID 12235
IUPAC navn 2,4-dimethylpentan-2-ol
SMIL CC(C)CC(C)(C)O

Specifications

Kemisk navn eller materiale 2,4-Dimethyl-2-pentanol
Tæthed 0.81
Kogepunkt 132°C to 134°C
Flammepunkt 38°C (100°F)
Brydningsindeks 1.417
Mængde 5 g
FN nummer UN1987
Beilstein 1731623
Formel vægt 116.2
Procent renhed 96%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.