Få mere at vide
2,4-Difluorobenzoyl Risperidone Impurity, TRC
Artikelnummer. 17961925
Skift visning
Mængde:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17961925 10 mg
17924545 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17961925 Leverandør Toronto Research Chemicals Leverandørnr. TRCD44571510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 158697-67-7
Molekylær formel C23 H27 F2 N3 O2
Molekylvægt (g/mol) 415.48
Synonym 4H-Pyrido[1,2-a]pyrimidin-4-one, 3-[2-[4-(2,4-difluorobenzoyl)-1-piperidinyl]ethyl]-6,7,8,9-tetrahydro-2-methyl-, 3-[2-[4-(2,4-Difluorobenzoyl)-1-piperidinyl]ethyl]-6,7,8,9-tetrahydro-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one, 3-[2-[4-(2,4-Difluorobenzoyl)piperidino]ethyl]-6,7,8,9-tetrahydro-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one, 3-[2-[4-(2,4-Difluorobenzoyl)piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one, Risperidone Imp. H (EP), Risperidone Difluoroketone
IUPAC navn 3-[2-[4-(2,4-difluorobenzoyl)piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-4-one
SMIL CC1=C(CCN2CCC(CC2)C(=O)c3ccc(F)cc3F)C(=O)N4CCCCC4=N1
Tekniske data
Kemisk navn eller materiale 3-[2-[4-(2,4-Difluorobenzoyl)piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one
Mængde 10 mg
InChI formel InChI=1S/C23H27F2N3O2/c1-15-18(23(30)28-10-3-2-4-21(28)26-15)9-13-27-11-7-16(8-12-27)22(29)19-6-5-17(24)14-20(19)25/h5-6,14,16H,2-4,7-13H2,1H3
Formel vægt 415.2071