Få mere at vide
2,4'-Dichlorobenzophenone, 98%, Thermo Scientific™
Artikelnummer. 11426750
Skift visning
Mængde:
5 g
25 g
Pakningsstørrelse:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11426750 5 g 5g
11436750 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 11426750 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. L07648.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 85-29-0
Molekylær formel C13H8Cl2O
Molekylvægt (g/mol) 251.11
MDL nummer MFCD00038744
InChI nøgle YXMYPHLWXBXNFF-UHFFFAOYSA-N
Synonym 2,4'-dichlorobenzophenone, 2-chlorophenyl 4-chlorophenyl methanone, methanone, 2-chlorophenyl 4-chlorophenyl, 2-chlorophenyl-4-chlorophenyl methanone, unii-h3n0v78i65, benzophenone, 2,4'-dichloro, di2-chlorophenyl ketone, pubchem3373, o,p-dichlorobenzophenone, o,p'-dichlorobenzophenone
PubChem CID 66558
IUPAC navn (2-chlorophenyl)-(4-chlorophenyl)methanone
SMIL ClC1=CC=C(C=C1)C(=O)C1=CC=CC=C1Cl
Tekniske data
Kemisk navn eller materiale 2,4'-Dichlorobenzophenone
Smeltepunkt 63°C to 66°C
Mængde 5 g
Beilstein 1959090
Formel vægt 251.11
Procent renhed 98%