Få mere at vide
2,4-Dichloro-beta-[(2,4-dichlorophenyl)methoxy]benzeneethanamine, TRC
Artikelnummer. 30133128
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133128 250 mg
30006281 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30133128 Leverandør Toronto Research Chemicals Leverandørnr. TRCD434120250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 67358-54-7
Molekylær formel C15 H13 Cl4 N O
Molekylvægt (g/mol) 365.08
Synonym 2,4-Dichloro-β-[(2,4-dichlorophenyl)methoxy]benzeneethanamine
IUPAC navn 2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethanamine
SMIL ClC1=CC=C(C(Cl)=C1)COC(C2=CC=C(Cl)C=C2Cl)CN
Tekniske data
Kemisk navn eller materiale 2,4-Dichloro-beta-[(2,4-dichlorophenyl)methoxy]benzeneethanamine
Renhedsgrad noter HPLC
Mængde 250 mg
InChI formel InChI=1S/C15H13Cl4NO/c16-10-2-1-9(13(18)5-10)8-21-15(7-20)12-4-3-11(17)6-14(12)19/h1-6,15H,7-8,20H2
Formel vægt 362.9751
Procent renhed >95%