Få mere at vide
2-(4-Chloro-3-sulfobenzoyl)-benzoic Acid, TRC
Artikelnummer. 30135170
Skift visning
Mængde:
10 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135170 100 mg
30133231 250 mg
30009057 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30135170 Leverandør Toronto Research Chemicals Leverandørnr. TRCC427810100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 345930-32-7
Molekylær formel C14 H9 Cl O6 S
Molekylvægt (g/mol) 340.74
Synonym Benzoic acid, 2-(4-chloro-3-sulfobenzoyl)-, 2-(4-Chloro-3-sulfobenzoyl)benzoic acid
IUPAC navn 2-(4-chloro-3-sulfobenzoyl)benzoic acid
SMIL OC(=O)c1ccccc1C(=O)c2ccc(Cl)c(c2)S(=O)(=O)O
Tekniske data
Kemisk navn eller materiale 2-(4-Chloro-3-sulfobenzoyl)-benzoic Acid
Renhedsgrad noter HPLC
Mængde 100 mg
InChI formel InChI=1S/C14H9ClO6S/c15-11-6-5-8(7-12(11)22(19,20)21)13(16)9-3-1-2-4-10(9)14(17)18/h1-7H,(H,17,18)(H,19,20,21)
Formel vægt 339.9808
Procent renhed >95%