Få mere at vide
(+)-2,3-O-Isopropylidene-L-threitol, 98%, Thermo Scientific™
Artikelnummer. 11446286
Skift visning
Mængde:
1 g
5 g
Pakningsstørrelse:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11446286 1 g 1g
11456286 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 11446286 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. B23090.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
(+)-2,3-O-Isopropylidene-L-threitol is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
Solubility
Soluble in water.
Notes
Hygroscopic. Store away from oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 50622-09-8
Molekylær formel C7H14O4
Molekylvægt (g/mol) 162.185
MDL nummer MFCD00063761
InChI nøgle INVRLGIKFANLFP-WDSKDSINSA-N
Synonym +-2,3-o-isopropylidene-l-threitol, 4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-diyl dimethanol, 2,3-o-isopropylidene-l-threitol, 4s,5s-5-hydroxymethyl-2,2-dimethyl-1,3-dioxolan-4-yl methanol, +-2,3,o-isopropylidene-l-threitol, pubchem19000, 2,3-o-isoproylidene-l-threitol, 1,3-dioxolane-4,5-dimethanol, 2,2-dimethyl-, 4s,5s, 2-o,3-o-isopropylidene-l-threitol, oi-2,3-o-isopropylidene-l-threitol
PubChem CID 785242
IUPAC navn [(4S,5S)-5-(hydroxymethyl)-2,2-dimethyl-1,3-dioxolan-4-yl]methanol
SMIL CC1(OC(C(O1)CO)CO)C
Tekniske data
Kemisk navn eller materiale (+)-2,3-O-Isopropylidene-L-threitol
Smeltepunkt 45°C to 49°C
Kogepunkt 92°C to 94°C (0.1 mmHg)
Flammepunkt >110°C (230°F)
Mængde 1 g
Beilstein 1280797
Følsomhed Hygroscopic
Opløselighedsinformation Soluble in water.
Optisk rotation +3° (c=5 in Ethanol)
Formel vægt 162.19
Procent renhed 98%