Få mere at vide
(2,3-Dimethylphenyl)-1H-imidazol-5-ylmethanone, TRC
Artikelnummer. 30012587
Skift visning
Mængde:
50 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30012587 250 mg
30008754 1 g
30008235 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30012587 Leverandør Toronto Research Chemicals Leverandørnr. TRCD480780250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 91874-85-0
Molekylær formel C12H12N2O
Molekylvægt (g/mol) 200.24
Synonym (2,3-dimethylphenyl)-1H-imidazol-4-ylmethanone
SMIL Cc1cccc(C(=O)c2cnc[nH]2)c1C
Tekniske data
Kemisk navn eller materiale (2,3-Dimethylphenyl)-1H-imidazol-5-ylmethanone
Mængde 250 mg
InChI formel InChI=1S/C12H12N2O/c1-8-4-3-5-10(9(8)2)12(15)11-6-13-7-14-11/h3-7H,1-2H3,(H,13,14)
Formel vægt 200.095