Få mere at vide
2,3,4-Trimethoxy-6-methylbenzaldehyde, TRC
Artikelnummer. 30456078
Skift visning
Mængde:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30456078 5 g
30449892 1 g
30452822 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30456078 Leverandør Toronto Research Chemicals Leverandørnr. TRCT7962355G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 22383-85-3
Molekylær formel C11H14O4
Molekylvægt (g/mol) 210.23
Synonym 2,3,4-Trimethoxy-6-methylbenzaldehyde, 2-Methyl-4,5,6-trimethoxybenzaldehyde, 4,5,6-Trimethoxy-o-tolualdehyde
IUPAC navn 2,3,4-trimethoxy-6-methylbenzaldehyde
SMIL COc1cc(C)c(C=O)c(OC)c1OC
Tekniske data
Kemisk navn eller materiale 2,3,4-Trimethoxy-6-methylbenzaldehyde
Mængde 5 g
InChI formel InChI=1S/C11H14O4/c1-7-5-9(13-2)11(15-4)10(14-3)8(7)6-12/h5-6H,1-4H3
Formel vægt 210.0892