Få mere at vide
2-(3,4-Dimethylphenyl)-1,2-dihydro-5-methyl-3H-pyrazol-3-one, TRC
Artikelnummer. 30133711
Skift visning
Mængde:
1 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133711 25 g
30136215 10 g
30012276 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133711 Leverandør Toronto Research Chemicals Leverandørnr. TRCD47693025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 277299-70-4
Molekylær formel C12 H14 N2 O
Molekylvægt (g/mol) 202.25
Synonym 3H-Pyrazol-3-one, 2-(3,4-dimethylphenyl)-1,2-dihydro-5-methyl-, 2-(3,4-Dimethylphenyl)-1,2-dihydro-5-methyl-3H-pyrazol-3-one, 1-(3,4-Dimethylphenyl)-3-methyl-3-pyrazolin-5-one
IUPAC navn 2-(3,4-dimethylphenyl)-5-methyl-1H-pyrazol-3-one
SMIL CC1=CC(=O)N(N1)c2ccc(C)c(C)c2
Tekniske data
Kemisk navn eller materiale 2-(3,4-Dimethylphenyl)-1,2-dihydro-5-methyl-3H-pyrazol-3-one
Mængde 25 g
InChI formel InChI=1S/C12H14N2O/c1-8-4-5-11(6-9(8)2)14-12(15)7-10(3)13-14/h4-7,13H,1-3H3
Formel vægt 202.1106